Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyphenylacetic acid, 98%

Catalog No. AAB2062522
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2062522 100 g
AAB2062514 25 g
AAB2062536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2062522 Supplier Thermo Scientific Chemicals Supplier No. B2062522
Add to Cart

CAS: 93-40-3 | C10H12O4 | 196.202 g/mol

It reacts with formaldehyde in the presence of acid to give an isochromanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It reacts with formaldehyde in the presence of acid to give an isochromanone.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-40-3
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00004335
InChI Key WUAXWQRULBZETB-UHFFFAOYSA-N
Synonym homoveratric acid, 3,4-dimethoxyphenylacetic acid, 3,4-dimethoxyphenyl acetic acid, 2-3,4-dimethoxyphenyl acetic acid, benzeneacetic acid, 3,4-dimethoxy, 3,4-dimethoxybenzeneacetic acid, 3,4-dimethoxy-phenyl-acetic acid, acetic acid, 3,4-dimethoxyphenyl
PubChem CID 7139
ChEBI CHEBI:86655
IUPAC Name 2-(3,4-dimethoxyphenyl)acetic acid
SMILES COC1=C(C=C(C=C1)CC(=O)O)OC

Specifications

Melting Point 95°C to 98°C
Quantity 100 g
Beilstein 1110282
Solubility Information Soluble in water.
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxyphenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-244-5
RTECSNumber AH0675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.