Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dimethylacetophenone, 98%

Catalog No. p-7081972
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 3637-01-2 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3637-01-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008743
InChI Key WPRAXAOJIODQJR-UHFFFAOYSA-N

Specifications

Density 1.002
Boiling Point 242°C to 243°C
Flash Point 102°C (215°F)
Refractive Index 1.538
Quantity 5 g
Beilstein 907053
Synonym 3',4'-dimethylacetophenone, 1-3,4-dimethylphenyl ethanone, 3,4-dimethylacetophenone, ethanone, 1-3,4-dimethylphenyl, 1-3,4-dimethylphenyl ethan-1-one, unii-pc1l145t5v, 1-3,4-dimethyl-phenyl-ethanone, 1-acetyl-3,4-dimethylbenzene, 3.4-dimethylacetophenon, acmc-1cmy2
SMILES CC1=C(C=C(C=C1)C(=O)C)C
IUPAC Name 1-(3,4-dimethylphenyl)ethanone
PubChem CID 77193
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 3',4'-Dimethylacetophenone
Show More Show Less
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 222-859-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.