Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylanisole, 99%

Catalog No. p-7026637
Click to view available options
Quantity:
50 g
250 g

CAS: 4685-47-6 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4685-47-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008396
InChI Key LVUBSVWMOWKPDJ-UHFFFAOYSA-N
Synonym 3,4-dimethylanisole, 4-methoxy-1,2-dimethyl-benzene, 4-methoxy-o-xylene, benzene, 4-methoxy-1,2-dimethyl, 3,4-dimethyl anisole, 1,2-dimethyl-4-methoxybenzene, 3,4-dimethylphenyl methyl ether, acmc-20anel, pubchem5361
PubChem CID 78411
IUPAC Name 4-methoxy-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)OC)C

Specifications

Density 0.975
Boiling Point 200°C to 201°C
Quantity 50 g
Refractive Index 1.5175
Beilstein 2040907
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3,4-Dimethylanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 225-142-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.