Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Formylphenyl)propionic acid, 96%

Catalog No. AAH3387306
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAH3387306

Supplier: Thermo Scientific Chemicals H3387306

Only null left
Add to Cart

CAS: 34961-64-3 | C10H10O3 | 178.19 g/mol

3-(4-Formylphenyl)propionic acid is used as an intermediate in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Formylphenyl)propionic acid is used as an intermediate in the manufacture of pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34961-64-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD08234824
InChI Key NRCCSDVEUWXOMG-UHFFFAOYSA-N
Synonym 3-4-formylphenyl propanoic acid, 3-4-formylphenyl propionic acid, 3-4-formyl-phenyl-propionic acid, 3-4-formylphenyl-propionic acid, 4-formyldihydrocinnamic acid, 4-formylbenzenepropanoic acid, benzenepropanoic acid,4-formyl, 3-4-formylphenyl propanoicacid
PubChem CID 19825241
IUPAC Name 3-(4-formylphenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=O)C=C1

Specifications

Melting Point ∼136°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 178.19
Percent Purity 96%
Chemical Name or Material 3-(4-Formylphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.