Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methylbenzoyl)propionic acid, 98%

Catalog No. AAA1029014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1029014 25 g
AAA1029006 5 g
AAA1029022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1029014 Supplier Thermo Scientific Chemicals Supplier No. A1029014
Add to Cart

CAS: 4619-20-9 | C11H12O3 | 192.214 g/mol

3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field

Solubility
Insoluble in water.

Notes
Incompatible with bases and oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4619-20-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00020541
InChI Key OEEUWZITKKSXAZ-UHFFFAOYSA-N
Synonym 3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo
PubChem CID 244162
IUPAC Name 4-(4-methylphenyl)-4-oxobutanoic acid
SMILES CC1=CC=C(C=C1)C(=O)CCC(=O)O

Specifications

Melting Point 124°C to 128°C
Quantity 25 g
Beilstein 2099653
Solubility Information Insoluble in water.
Formula Weight 192.22
Percent Purity 98%
Chemical Name or Material 3- (4-Methylbenzoyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.