Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-(Methylenedioxy)benzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
94839-07-3
C7H7BO4
165.939
MFCD01009695
CMHPUBKZZPSUIQ-UHFFFAOYSA-N
3,4-methylenedioxyphenylboronic acid,benzo d 1,3 dioxol-5-ylboronic acid,3,4-methylenedioxy phenylboronic acid,3,4-methylenedioxyphenyl boronic acid,3,4-methylenedioxy benzeneboronic acid,1,3-benzodioxole-5-boronic acid,3,4-methylenedioxybenzeneboronic acid,2h-1,3-benzodioxol-5-yl boronic acid,2h-1,3-benzodioxol-5-yl-boranediol,2h-1,3-benzodioxol-5-ylboronic acid
2734371
1,3-benzodioxol-5-ylboronic acid
B(C1=CC2=C(C=C1)OCO2)(O)O
94839-07-3
C7H7BO4
165.939
MFCD01009695
CMHPUBKZZPSUIQ-UHFFFAOYSA-N
3,4-methylenedioxyphenylboronic acid,benzo d 1,3 dioxol-5-ylboronic acid,3,4-methylenedioxy phenylboronic acid,3,4-methylenedioxyphenyl boronic acid,3,4-methylenedioxy benzeneboronic acid,1,3-benzodioxole-5-boronic acid,3,4-methylenedioxybenzeneboronic acid,2h-1,3-benzodioxol-5-yl boronic acid,2h-1,3-benzodioxol-5-yl-boranediol,2h-1,3-benzodioxol-5-ylboronic acid
2734371
1,3-benzodioxol-5-ylboronic acid
B(C1=CC2=C(C=C1)OCO2)(O)O
Melting Point 239°C to 243°C
Quantity 1g
Beilstein 5523347
Formula Weight 165.94
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning