Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Piperidyl)indole 98.0+%, TCI America™
SDP

Catalog No. P23221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23221G 1 g
P2322200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23221G Supplier TCI America Supplier No. P23221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17403-09-7
Molecular Formula C13H17N2
Molecular Weight (g/mol) 201.29
MDL Number MFCD01764670
InChI Key KAIRZPVWWIMPFT-UHFFFAOYSA-O
Synonym 3-piperidin-4-yl-1h-indole, 3-4-piperidyl-1h-indole, 3-4-piperidyl indole, 3-4-piperidinyl-1h-indole, 4-3-indolyl piperidine, 4-3-indo piperidine, 3-piperid-4-yl indole, 4-1h-indol-3-yl piperidine, 1h-indole,3-4-piperidinyl, peakdale1_002265
PubChem CID 702235
IUPAC Name 4-(1H-indol-3-yl)piperidin-1-ium
SMILES C1CC(CC[NH2+]1)C1=CNC2=CC=CC=C12

Specifications

Melting Point 223°C
Color Yellow
Quantity 1 g
Formula Weight 200.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Piperidyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.