Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-tert-Butylphenyl)isobutyraldehyde 96.0+%, TCI America™
SDP

Catalog No. B1145100ML
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Chemical Identifiers

CAS 80-54-6
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00047655
InChI Key SDQFDHOLCGWZPU-UHFFFAOYNA-N
Synonym lilial, 3-4-tert-butyl phenyl-2-methylpropanal, 3-4-tert-butylphenyl isobutyraldehyde, lilyal, lily aldehyde, 3-4-tert-butylphenyl-2-methylpropanal, 3-4-tert-butylphenyl-2-methylpropionaldehyde, benzenepropanal, 4-1,1-dimethylethyl-.alpha.-methyl, butylphenyl methylpropional, 2-4-tert-butylbenzyl propionaldehyde
PubChem CID 228987
IUPAC Name 3-(4-tert-butylphenyl)-2-methylpropanal
SMILES CC(CC1=CC=C(C=C1)C(C)(C)C)C=O

Specifications

Color Yellow
Boiling Point 150°C
Quantity 100 mL
Formula Weight 204.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(4-tert-Butylphenyl)isobutyraldehyde
EINECSNumber (3)-2667
RTECSNumber MW4895000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.