Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5,6,8-Tetrabromo-1,10-phenanthroline 95.0+%, TCI America™
SDP

Catalog No. T3133100MG
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 66127-00-2
Molecular Formula C12H4Br4N2
Molecular Weight (g/mol) 495.79
MDL Number MFCD09879848
InChI Key BAPJVWLAFSQLNX-UHFFFAOYSA-N
PubChem CID 11443337
IUPAC Name 3,5,6,8-tetrabromo-1,10-phenanthroline
SMILES BrC1=CN=C2C(=C1)C(Br)=C(Br)C1=CC(Br)=CN=C21

Specifications

Melting Point 356°C
Color White-Yellow
Quantity 100 mg
Formula Weight 495.79
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5,6,8-Tetrabromo-1,10-phenanthroline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.