Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)aniline, 98+%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
328-74-5
C8H5F6N
229.12
MFCD00000394
CDIDGWDGQGVCIB-UHFFFAOYSA-N
3,5-bis trifluoromethyl aniline,3,5-di trifluoromethyl aniline,benzenamine, 3,5-bis trifluoromethyl,3,5-bis trifluoromethyl benzenamine,3',5'-bis-trifluoromethyl aniline,3,5-bis-trifluoromethylaniline,alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine,3,5-bis-trifluoromethyl-phenylamine,aniline, 3,5-bis trifluoromethyl
9480
3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F
328-74-5
C8H5F6N
229.12
MFCD00000394
CDIDGWDGQGVCIB-UHFFFAOYSA-N
3,5-bis trifluoromethyl aniline,3,5-di trifluoromethyl aniline,benzenamine, 3,5-bis trifluoromethyl,3,5-bis trifluoromethyl benzenamine,3',5'-bis-trifluoromethyl aniline,3,5-bis-trifluoromethylaniline,alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine,3,5-bis-trifluoromethyl-phenylamine,aniline, 3,5-bis trifluoromethyl
9480
3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F
Specific Gravity 1.47
Density 1.4700g/mL
Boiling Point 85°C (15.0mmHg)
Flash Point 83°C
Refractive Index 1.4330 to 1.435
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50mL
Color Brown-Yellow to Yellow
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CF3)2C6H3NH2
Formula Weight 229.12
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 3, 5-Bis(trifluoromethyl)aniline
Show More Show Less

GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remov

GHS Signal Word: Warning

EINECSNumber : 206-335-