Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)benzaldehyde 95.0+%, TCI America™
SDP

Catalog No. B175125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B175125G 25 g
B17515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B175125G Supplier TCI America Supplier No. B175125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 401-95-6
Molecular Formula C9H4F6O
Molecular Weight (g/mol) 242.12
MDL Number MFCD00010206
InChI Key LDWLIXZSDPXYDR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
PubChem CID 136247
IUPAC Name 3,5-bis(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O

Specifications

Color Yellow
Boiling Point 171°C
Quantity 25 g
Formula Weight 242.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.