Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Catalog No. AA4345203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4345203 1 g
AA4345206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4345203 Supplier Thermo Scientific Chemicals Supplier No. 4345203

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-29-7
Molecular Formula C9H7F6N
Molecular Weight (g/mol) 243.152
MDL Number MFCD00009909
InChI Key DHVHORCFFOSRBP-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
PubChem CID 521099
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]methanamine
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN

Specifications

Melting Point 50°C to 55°C
Flash Point 78°C (173°F)
Quantity 1 g
UN Number UN3259
Formula Weight 243.15
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.