Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Diacetoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. D197810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D197810G 10 g
D197825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D197810G Supplier TCI America Supplier No. D197810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35086-59-0
Molecular Formula C12H12O5
Molecular Weight (g/mol) 236.223
MDL Number MFCD00008702
InChI Key QODJHYBESCIPOG-UHFFFAOYSA-N
PubChem CID 688015
IUPAC Name (3-acetyl-5-acetyloxyphenyl) acetate
SMILES CC(=O)C1=CC(=CC(=C1)OC(=O)C)OC(=O)C

Specifications

Melting Point 93°C
Quantity 10 g
Formula Weight 236.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Diacetoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.