Learn More
3,5-Diamino-1,2,4-triazole, 98%
Description
3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water.
Notes
Air sensitive. Incompatible with acids and oxidizing agents.
Specifications
Specifications
| Melting Point | ∼205°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 112467 |
| Solubility Information | Soluble in water. |
| Formula Weight | 99.1 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 3,5-Diamino-1,2,4-triazole |
Safety and Handling
| EINECSNumber | 215-937-2 |
| RTECSNumber | XZ4535000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.