Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diaminobenzoic acid, 98%

Catalog No. p-100515
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

Catalog No. AC112111000

Supplier: Thermo Scientific Chemicals 112111000

Only null left
Add to Cart

CAS: 535-87-5 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-87-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007807
InChI Key UENRXLSRMCSUSN-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
PubChem CID 12062
IUPAC Name 3,5-diaminobenzoic acid
SMILES C1=C(C=C(C=C1N)N)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 235°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (H2N)2C6H3CO2H
Quantity 100 g
Beilstein 14, 453
Merck Index 15, 2980
Solubility Information Solubility in water: 7g/L (22°C). Other solubilities: soluble in alcohol and ether
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 5-Diaminobenzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-621-0
RTECSNumber DG6186000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.