Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture

Catalog No. aaa1407518
Click to view available options
Quantity:
50 g
100 g
500 g

Catalog No. AAA1407518

Supplier: Thermo Scientific Chemicals A1407518

Only null left
Add to Cart

CAS: 535-87-5 | C7H8N2O2 | 152.153 g/mol

3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-87-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007807
InChI Key UENRXLSRMCSUSN-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
PubChem CID 12062
IUPAC Name 3,5-diaminobenzoic acid
SMILES C1=C(C=C(C=C1N)N)C(=O)O

Specifications

Melting Point ∼235°C (decomposition)
Flash Point 210°C (410°F)
Quantity 50 g
Beilstein 2086484
Merck Index 14,2978
Solubility Information Very soluble in water.
Formula Weight 152.15
Percent Purity 98%
Assay may contain up to 3% moisture
Chemical Name or Material 3,5-Diaminobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-621-0
RTECSNumber DG6186000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.