Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibenzyloxybenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D265125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D265125G 25 g
D26515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D265125G Supplier TCI America Supplier No. D265125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24131-31-5
Molecular Formula C21H20O3
Molecular Weight (g/mol) 320.388
MDL Number MFCD01863236
InChI Key MHHXKZKHZMSINU-UHFFFAOYSA-N
Synonym 3,5-dibenzyloxybenzyl alcohol, 3,5-bis benzyloxy phenyl methanol, 3,5-bis benzyloxy benzyl alcohol, 3,5-bis phenylmethoxy phenyl methanol, 3,5-benzyloxybenzyl alcohol, 3,5-bis phenylmethoxy phenyl methan-1-ol, pubchem13641, 3,5-dibenzyloxyphenyl methanol, mhhxkzkhzmsinu-uhfffaoysa
PubChem CID 561476
IUPAC Name [3,5-bis(phenylmethoxy)phenyl]methanol
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)CO)OCC3=CC=CC=C3

Specifications

Melting Point 82°C
Color White
Quantity 25 g
Formula Weight 320.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.