Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibenzyloxybenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. B209325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B209325G 25 g
B20935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B209325G Supplier TCI America Supplier No. B209325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24131-32-6
Molecular Formula C21H19BrO2
Molecular Weight (g/mol) 383.285
MDL Number MFCD02093444
InChI Key WGMYJGAUAQXYFQ-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzyl Bromide, 3,5-Dibenzyloxy-alpha-bromotoluene
PubChem CID 2761019
IUPAC Name 1-(bromomethyl)-3,5-bis(phenylmethoxy)benzene
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)CBr)OCC3=CC=CC=C3

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 383.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.