Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromo-o-phenylenediamine monohydrochloride, 99%, Thermo Scientific™

Catalog No. ac223731000
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle

Chemical Identifiers

CAS 75568-11-5
Molecular Formula C6H6Br2N2·HCl
Molecular Weight (g/mol) 302.4
MDL Number MFCD00012967
InChI Key QOZDVKFQMGARCC-UHFFFAOYSA-N
Synonym 3,5-dibromo-1,2-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine hydrochloride, acmc-20andn, 1,2-diamino-3,5-dibromobenzene monohydrochloride, 3,5-bis bromanyl benzene-1,2-diamine hydrochloride, 3,5-dibromo-1,2-phenylenediamine hydrochloride, 1,2-benzenediamine,3,5-dibromo-, hydrochloride 1:1, 3,5-dibromo-ortho-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine-hydrogen chloride 1/1, 3,5-dibromo-1,2-phenylenediaminemonohydrochloride sensitivereagentforthedeterminationofsebygc-ecd
PubChem CID 2724285
IUPAC Name 3,5-dibromobenzene-1,2-diamine;hydrochloride
SMILES C1=C(C=C(C(=C1Br)N)N)Br.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 238°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Br2C6H2(NH2)2·HCl
Quantity 100 mg
Beilstein 13, 29
Formula Weight 302.4
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 5-Dibromo-o-phenylenediamine monohydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.