Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromoanisole 98.0+%, TCI America™
SDP

Catalog No. D336425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D336425G 25 g
D33645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D336425G Supplier TCI America Supplier No. D336425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74137-36-3
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD02258848
InChI Key OQZAQBGJENJMHT-UHFFFAOYSA-N
Synonym 3,5-dibromoanisole, 1,3-dibromo-5-methoxy-benzene, benzene, 1,3-dibromo-5-methoxy, pubchem2009, acmc-1bh5r, ksc493q1d, 1.3-dibromo-5-methoxybenzene
PubChem CID 11021812
IUPAC Name 1,3-dibromo-5-methoxybenzene
SMILES COC1=CC(=CC(=C1)Br)Br

Specifications

Melting Point 39°C
Boiling Point 261°C
Quantity 25 g
Formula Weight 265.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromoanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.