Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,5-Dibromobenzoic acid, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
Specifications
Melting Point | 212.0°C to 214.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass Bottle |
Linear Formula | Br2C6H3CO2H |
Quantity | 1 g |
Formula Weight | 279.91 |
Percent Purity | 99% |
Chemical Name or Material | 3,5-Dibromobenzoic acid |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction