Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromopyridine N-Oxide 98.0+%, TCI America™
SDP

Catalog No. D43261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43261G 1 g
D43265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43261G Supplier TCI America Supplier No. D43261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2402-99-5
Molecular Formula C5H3Br2NO
Molecular Weight (g/mol) 252.893
MDL Number MFCD00160750
InChI Key QBUQQUOGXPOBAQ-UHFFFAOYSA-N
PubChem CID 620003
IUPAC Name 3,5-dibromo-1-oxidopyridin-1-ium
SMILES C1=C(C=[N+](C=C1Br)[O-])Br

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 1 g
Synonym 3,5-dibromopyridine 1-oxide, 3,5-dibromopyridine n-oxide, 3,5-dibromopyridine-n-oxide, 3,5-dibromopyridin-1-ium-1-olate, 3,5-dibromopyridine1-oxide, 3,5-dibromopyridine-1-oxide, pyridine, 3,5-dibromo-,1-oxide, pubchem6616, 3,5-dibromopyridin-1-ol, 3,5-dibromopyridine n oxide
Formula Weight 252.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromopyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.