Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2,4-difluoroaniline 94.0+%, TCI America™
SDP

Catalog No. D363725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D363725G 25 g
D36375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D363725G Supplier TCI America Supplier No. D363725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83121-15-7
Molecular Formula C6H3Cl2F2N
Molecular Weight (g/mol) 197.99
MDL Number MFCD00044104
InChI Key KLECNQGLBJHVSH-UHFFFAOYSA-N
PubChem CID 2774000
IUPAC Name 3,5-dichloro-2,4-difluoroaniline
SMILES NC1=CC(Cl)=C(F)C(Cl)=C1F

Specifications

Melting Point 75°C
Quantity 25 g
Formula Weight 197.99
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-2,4-difluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.