Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%

Catalog No. aaa1073209
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AAA1073209

Supplier: Thermo Scientific Chemicals A1073209

Only null left
Add to Cart

CAS: 54970-72-8 | C6H3Cl2NaO4S | 265.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54970-72-8
Molecular Formula C6H3Cl2NaO4S
Molecular Weight (g/mol) 265.036
MDL Number MFCD00009798
InChI Key NMWCVZCSJHJYFW-UHFFFAOYSA-M
Synonym sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
PubChem CID 3085077
IUPAC Name sodium;3,5-dichloro-2-hydroxybenzenesulfonate
SMILES C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 3640643
Formula Weight 265.05
Percent Purity 99%
Chemical Name or Material 3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 259-416-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.