Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. D326125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D326125G 25 g
D32615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D326125G Supplier TCI America Supplier No. D326125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6575-00-4
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.01
MDL Number MFCD00001800
InChI Key PUJSUOGJGIECFQ-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-dichloro, 3,5-dichloro-benzonitrile, 3,5-dichlorobenzenecarbonitrile, pubchem4752, 3,5-dichlorobenzonitril, acmc-1b8n0, ksc354e0l, chembl51500, 3,5-dichlorobenzonitrile
PubChem CID 81052
IUPAC Name 3,5-dichlorobenzonitrile
SMILES ClC1=CC(=CC(Cl)=C1)C#N

Specifications

Melting Point 68°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 172.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.