Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D379125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D379125G 25 g
D37915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D379125G Supplier TCI America Supplier No. D379125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60211-57-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004634
InChI Key VSNNLLQKDRCKCB-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
PubChem CID 43236
IUPAC Name (3,5-dichlorophenyl)methanol
SMILES C1=C(C=C(C=C1Cl)Cl)CO

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Formula Weight 177.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.