Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorophenylhydrazine, 98%, Thermo Scientific™

Catalog No. AAA1120703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1120703 1 g
AAA1120706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1120703 Supplier Thermo Scientific Chemicals Supplier No. A1120703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39943-56-1
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.028
MDL Number MFCD00051527
InChI Key ZQOCIFROLPHOEF-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl hydrazine, 3,5-dichloro phenylhydrazine, hydrazine, 3,5-dichlorophenyl, 3,5-dichlorophenylhydrazinehydrochloride, acmc-1cttv, 3,5-dichloroaniline amine, zqocifrolphoef-uhfffaoysa, 3,5-dichloro-phenyl-hydrazine, 1-3,5-dichlorophenyl hydrazine
PubChem CID 600469
IUPAC Name (3,5-dichlorophenyl)hydrazine
SMILES C1=C(C=C(C=C1Cl)Cl)NN

Specifications

Melting Point 112°C to 117°C
Quantity 1 g
Beilstein 3126965
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 3,5-Dichlorophenylhydrazine
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.