Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorophenylhydrazine Hydrochloride 90.0+%, TCI America™
SDP

Catalog No. p-7127421
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 63352-99-8
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012938
InChI Key BDXZOJVMTJOAPS-UHFFFAOYSA-N
Synonym 3,5-dichlorophenylhydrazine hydrochloride, 3,5-dichlorophenyl hydrazine hydrochloride, 3,5-dichlorophenyl hydrazine monohydrochloride, 3,5-dichlorophenylhydrazine hcl, 1-3,5-dichlorophenyl hydrazine hydrochloride, hydrazine, 3,5-dichlorophenyl-, monohydrochloride, 3,5-dichlorophenylhydrazinehydrochloride, pubchem7531, c6h6cl2n2.hcl, acmc-209nf3
PubChem CID 2723913
IUPAC Name (3,5-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1Cl)Cl)NN.Cl

Specifications

Quantity 5 g
Formula Weight 213.49
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0472
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.