Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorosalicylaldehyde, 98%

Catalog No. AAB2094114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2094114 25 g
AAB2094106 5 g
AAB2094122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2094114 Supplier Thermo Scientific Chemicals Supplier No. B2094114
Add to Cart

CAS: 90-60-8 | C7H4Cl2O2 | 191.007 g/mol

3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.

Solubility
Insoluble in water. Solubility in methanol is almost transparent.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-60-8
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00003320
InChI Key FABVMBDCVAJXMB-UHFFFAOYSA-N
Synonym 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro
PubChem CID 66660
IUPAC Name 3,5-dichloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Cl)O)C=O)Cl

Specifications

Melting Point 94°C to 97°C
Quantity 25 g
Beilstein 973391
Sensitivity Air Sensitive
Solubility Information Insoluble in water. Solubility in methanol is almost transparent.
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 3,5-Dichlorosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-005-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.