Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorosalicylaldehyde 98.0+%, TCI America™
SDP

Catalog No. D117025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D117025G 25 g
D11705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D117025G Supplier TCI America Supplier No. D117025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90-60-8
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00003320
InChI Key FABVMBDCVAJXMB-UHFFFAOYSA-N
Synonym 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro
PubChem CID 66660
IUPAC Name 3,5-dichloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Cl)O)C=O)Cl

Specifications

Melting Point 94°C
Quantity 25 g
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorosalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.