Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorothioanisole 98.0+%, TCI America™
SDP

Catalog No. D352025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D352025G 25 g
D35205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D352025G Supplier TCI America Supplier No. D352025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68121-46-0
Molecular Formula C7H6Cl2S
Molecular Weight (g/mol) 193.09
MDL Number MFCD00870149
InChI Key PSSCMQMQOXVMFG-UHFFFAOYSA-N
Synonym 1,3-Dichloro-5-(methylthio)benzene
PubChem CID 129278
IUPAC Name 1,3-dichloro-5-(methylsulfanyl)benzene
SMILES CSC1=CC(Cl)=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 g
Formula Weight 193.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dichlorothioanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.