Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine 98.0+%, TCI America™
SDP

Catalog No. D055825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D055825G 25 g
D05581G 1 g
D05585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D055825G Supplier TCI America Supplier No. D055825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 632-93-9
Molecular Formula C14H21NO4
Molecular Weight (g/mol) 267.33
MDL Number MFCD00005950
InChI Key CDVAIHNNWWJFJW-UHFFFAOYSA-N
Synonym ddc porphyrinogen, 3,5-diethoxycarbonyl-1,4-dihydro-2,4,6-collidine, dicarbethoxydihydrocollidine, 3,5-diethoxycarbonyl-1,4-dihydrocollidine, 3,5-dicarbethoxy-1,4-dihydrocollidine, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,4,6-trimethyl-, diethyl ester, ddc van, 3,5-bis ethoxycarbonyl-1,4-dihydrocollidine, 1,4-dihydro-3,5-dicarbethoxycollidine, diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate
PubChem CID 12446
ChEBI CHEBI:83605
IUPAC Name 3,5-diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(C)NC(C)=C(C1C)C(=O)OCC

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 25 g
Formula Weight 267.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine
RTECSNumber US7979375
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.