Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Difluoroacetophenone 97.0+%, TCI America™
SDP

Catalog No. D342525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D342525G 25 g
D34255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D342525G Supplier TCI America Supplier No. D342525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123577-99-1
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00042489
InChI Key OXJLDNSPGPBDCP-UHFFFAOYSA-N
Synonym 3',5'-difluoroacetophenone, 3,5-difluoroacetophenone, 1-3,5-difluorophenyl ethanone, 1-3,5-difluorophenyl ethan-1-one, ethanone, 1-3,5-difluorophenyl, 1-acetyl-3,5-difluorobenzene, 1-3,5-difluoro-phenyl-ethanone, pubchem2179, acmc-1cacd, 3,5-di-fluoroacetophenone
PubChem CID 518596
IUPAC Name 1-(3,5-difluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC(F)=CC(F)=C1

Specifications

Melting Point 40°C
Boiling Point 95°C
Quantity 25 g
Formula Weight 156.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Difluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.