Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,5-Difluorobenzamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
874880-59-8
C7H6F2N2O
172.135
MFCD06796654
GYTSCORJTXOMMI-UHFFFAOYSA-N
3,5-difluorobenzamidoxime,3,5-difluoro-n-hydroxy-benzamidine,3,5-difluorophenyl hydroxyimino methylamine,3,5-difluoro-n'-hydroxybenzene-1-carboximidamide,e-3,5-difluoro-n'-hydroxybenzenecarboximidamide
9652016
3,5-difluoro-N'-hydroxybenzenecarboximidamide
C1=C(C=C(C=C1F)F)C(=NO)N
874880-59-8
C7H6F2N2O
172.135
MFCD06796654
GYTSCORJTXOMMI-UHFFFAOYSA-N
3,5-difluorobenzamidoxime,3,5-difluoro-n-hydroxy-benzamidine,3,5-difluorophenyl hydroxyimino methylamine,3,5-difluoro-n'-hydroxybenzene-1-carboximidamide,e-3,5-difluoro-n'-hydroxybenzenecarboximidamide
9652016
3,5-difluoro-N'-hydroxybenzenecarboximidamide
C1=C(C=C(C=C1F)F)C(=NO)N
Quantity 250mg
Color White
Formula Weight 172.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3,5-Difluorobenzamidoxime