Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D259625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 455-40-3
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00010323
InChI Key GONAVIHGXFBTOZ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoicacid, benzoic acid, 3,5-difluoro, pubchem2173, 3,5 difluorobenzoic acid, 3,5-difluoro benzoicacid, acmc-209k3a, 3,5 difluoro benzoic acid, 3,5-di-fluorobenzoic acid, 3,5-difluoro-benzoic acid
PubChem CID 101265
IUPAC Name 3,5-difluorobenzoic acid
SMILES C1=C(C=C(C=C1F)F)C(=O)O

Specifications

Melting Point 122°C
Color Purple-White
Quantity 25 g
Formula Weight 158.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.