Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzyl alcohol, 97%, Thermo Scientific™

Catalog No. AC305760050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 79538-20-8
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010313
InChI Key LOGIHEKXJKHXEC-UHFFFAOYSA-N
Synonym 3,5-difluorobenzyl alcohol, 3,5-difluorophenyl methanol, benzenemethanol, 3,5-difluoro, 3,5-difluorophenyl methan-1-ol, 3,5-difluorobenzylalcohol, 3, 5-difluorobenzyl alcohol, pubchem4918, acmc-1baxh, ksc493g5j, 3,5-difluoro-phenyl methanol
PubChem CID 522721
IUPAC Name (3,5-difluorophenyl)methanol
SMILES C1=C(C=C(C=C1F)F)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Colorless
Density 1.27g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Linear Formula FC6H3CHOH
Quantity 5 g
Formula Weight 144.12
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3, 5-Difluorobenzyl alcohol, 97%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.