Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorosalicylic acid, 98+%, Thermo Scientific Chemicals

Catalog No. p-7031758
Click to view available options
Quantity:
1 g

Catalog No. AAH3294003

Supplier: Thermo Scientific Chemicals H3294003

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84376-20-5
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.103
MDL Number MFCD06203609
InChI Key GZPCNALAXFNOBT-UHFFFAOYSA-N
Synonym 3,5-difluorosalicylic acid, 3,5-difluoro-salicylic acid, benzoic acid, 3,5-difluoro-2-hydroxy
PubChem CID 10888405
IUPAC Name 3,5-difluoro-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1F)O)C(=O)O)F

Specifications

Melting Point 187°C to 191°C
Quantity 1 g
Formula Weight 174.1
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.