Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dihydroxytoluene, 99%

Catalog No. aal1856706
Click to view available options
Quantity:
5 g
25 g

CAS: 504-15-4 | C7H8O2 | 124.14 g/mol

Used in production of orcein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-15-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002291
InChI Key OIPPWFOQEKKFEE-UHFFFAOYSA-N
Synonym orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
PubChem CID 10436
ChEBI CHEBI:16536
IUPAC Name 5-methylbenzene-1,3-diol
SMILES CC1=CC(O)=CC(O)=C1

Specifications

Melting Point 106°C to 108°C
Boiling Point 287°C to 290°C
Quantity 5 g
Beilstein 1071903
Sensitivity Air Sensitive
Merck Index 14,6864
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 3,5-Dihydroxytoluene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-984-2
RTECSNumber VH2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.