Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodo-L-thyronine 96.0+%, TCI America™
SDP

Catalog No. D11191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11191G 1 g
D1119100MG 100 mg
D11195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D11191G Supplier TCI America Supplier No. D11191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1041-01-6
Molecular Formula C15H13I2NO4
Molecular Weight (g/mol) 525.081
MDL Number MFCD00064987
InChI Key ZHSOTLOTTDYIIK-ZDUSSCGKSA-N
PubChem CID 92859
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid
SMILES C1=CC(=CC=C1O)OC2=C(C=C(C=C2I)CC(C(=O)O)N)I

Specifications

Melting Point 256°C
Quantity 1 g
Formula Weight 525.08
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodo-L-thyronine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.