Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diisopropylpyrazole 98.0+%, TCI America™
SDP

Catalog No. D240725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D240725G 25 g
D24075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D240725G Supplier TCI America Supplier No. D240725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17536-00-4
Molecular Formula C9H16N2
Molecular Weight (g/mol) 152.241
MDL Number MFCD00671516
InChI Key PJSJVTZSFIRYIZ-UHFFFAOYSA-N
PubChem CID 5255229
IUPAC Name 3,5-di(propan-2-yl)-1H-pyrazole
SMILES CC(C)C1=CC(=NN1)C(C)C

Specifications

Melting Point 88°C
Color White
Quantity 25 g
Formula Weight 152.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diisopropylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.