Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%

Catalog No. AC115540010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 2478-38-8 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2478-38-8
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008748
InChI Key OJOBTAOGJIWAGB-UHFFFAOYSA-N
Synonym acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxy
PubChem CID 17198
ChEBI CHEBI:2404
IUPAC Name 1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone
SMILES COC1=CC(=CC(OC)=C1O)C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 123°C to 128°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOC6H2(OCH3)2COCH3
Quantity 1 g
Beilstein 08,II,444
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,acetone,ether
Formula Weight 196.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 219-610-5
RTECSNumber AM8440000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.