Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxybenzylamine 98.0+%, TCI America™
SDP

Catalog No. D402525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D402525G 25 g
D40255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D402525G Supplier TCI America Supplier No. D402525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34967-24-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052813
InChI Key YGZJTYCCONJJGZ-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine
PubChem CID 420973
IUPAC Name (3,5-dimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1)CN)OC

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 96°C
Quantity 25 g
UN Number 3259
Formula Weight 167.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dimethoxybenzylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.