Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxyphenol, 98%

Catalog No. AAB2228318
Click to view available options
Quantity:
10 g
50 g

CAS: 500-99-2 | C8H10O3 | 154.17 g/mol

3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.

Solubility
Soluble in methanol, ethanol, benzene, water (slightly), and chloroform.

Notes
Air and light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-99-2
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008388
InChI Key XQDNFAMOIPNVES-UHFFFAOYSA-N
Synonym taxicatigenin, phenol, 3,5-dimethoxy, phloroglucinol dimethyl ether, 1-hydroxy-3,5-dimethoxybenzene, 3,5-dimethoxy phenol, unii-23uxw8136a, phenol,5-dimethoxy, 3,5-dimethoxy-phenol, pubchem16421, acmc-209kk2
PubChem CID 10383
IUPAC Name 3,5-dimethoxyphenol
SMILES COC1=CC(OC)=CC(O)=C1

Specifications

Melting Point 45°C to 47°C
Boiling Point 172°C to 175°C (17 mmHg)
Flash Point 78°C (173°F)
Quantity 50 g
Beilstein 1239156
Sensitivity Air and light sensitive
Solubility Information Soluble in methanol,ethanol,benzene,water (slightly),and chloroform.
Formula Weight 154.16
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-917-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.