Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxytoluene, 99%, Thermo Scientific™

Catalog No. ac391190010
Click to view available options
:
1g; Glass bottle

Specifications

CAS 4179-19-5
CAS Min % 98.5
CAS Max % 100.0
Density 1.0390g/mL
Boiling Point 244.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O2
Refractive Index 1.5210 to 1.5230 (20°C, 589nm)
Linear Formula CH3C6H3(OCH3)2
Quantity 1g
MDL Number MFCD00015435
Specific Gravity 1.039
Synonym 3,5-dimethoxytoluene, orcinol dimethyl ether, benzene, 1,3-dimethoxy-5-methyl, 5-methylresorcinol dimethyl ether, 3,5-dimethoxy toluene, toluene, 3,5-dimethoxy, 1,3-dimethoxy-5-methyl-benzene, pubchem17775, acmc-1ak5i
InChI Key RIZBLVRXRWHLFA-UHFFFAOYSA-N
SMILES COC1=CC(OC)=CC(C)=C1
IUPAC Name 1,3-dimethoxy-5-methylbenzene
Molecular Weight (g/mol) 152.19
PubChem CID 77844
Formula Weight 152.19
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethoxytoluene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 224-048-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.