Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1-adamantanamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D360825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D360825G 25 g
D36085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D360825G Supplier TCI America Supplier No. D360825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41100-52-1
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00214336
InChI Key LDDHMLJTFXJGPI-UHFFFAOYNA-N
Synonym memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
PubChem CID 181458
ChEBI CHEBI:64323
IUPAC Name hydrogen 3,5-dimethyladamantan-1-amine chloride
SMILES [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2

Specifications

Melting Point 292°C
Color White
Quantity 25 g
Formula Weight 215.77
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-1-adamantanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.