Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1H-pyrazole, 99%

Catalog No. p-7080577
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 67-51-6 | C5H8N2 | 96.13 g/mol

3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI)­ oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI)­ oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.

Solubility
Soluble in water, methanol 0.1 g/mL.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-51-6
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005243
InChI Key SDXAWLJRERMRKF-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256
PubChem CID 6210
IUPAC Name 3,5-dimethyl-1H-pyrazole
SMILES CC1=CC(C)=NN1

Specifications

Melting Point 105°C to 108°C
pH 6.7
Boiling Point 218°C
Quantity 50 g
Beilstein 106325
Solubility Information Soluble in water,methanol 0.1g/mL.
Formula Weight 96.13
Percent Purity 99%
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole
Hazard Category H302-H361fd-H373
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-657-5
RTECSNumber UQ6477600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.