Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylbenzoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
499-06-9
C9H10O2
150.18
MFCD00002525
UMVOQQDNEYOJOK-UHFFFAOYSA-N
mesitylenic acid,benzoic acid, 3,5-dimethyl,unii-ed8av34n0y,3,5-dimethyl benzoic acid,3,5-dimethyl-benzoic acid,ed8av34n0y,3,5-dimethylbenzoicacid,pubchem15440,3,5-dimethylbezoic acid,3,5 dimethylbenzoic acid
10356
CHEBI:64821
3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1)C(=O)O)C
499-06-9
C9H10O2
150.18
MFCD00002525
UMVOQQDNEYOJOK-UHFFFAOYSA-N
mesitylenic acid,benzoic acid, 3,5-dimethyl,unii-ed8av34n0y,3,5-dimethyl benzoic acid,3,5-dimethyl-benzoic acid,ed8av34n0y,3,5-dimethylbenzoicacid,pubchem15440,3,5-dimethylbezoic acid,3,5 dimethylbenzoic acid
10356
CHEBI:64821
3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1)C(=O)O)C
Color White to Yellow
Melting Point 169°C to 172°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3CO2H
Beilstein 09, 536
Solubility Information Solubility in water: slightly soluble
Formula Weight 150.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 3, 5-Dimethylbenzoic acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-876-5

RTECSNumber : DG8734030

TSCA : TSCA