Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylisoxazole, 99%

Catalog No. AAA1221722
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAA1221722

Supplier: Thermo Scientific Chemicals A1221722

Only null left
Add to Cart

CAS: 300-87-8 | C5H7NO | 97.117 g/mol

The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-87-8
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.117
MDL Number MFCD00003156
InChI Key FICAQKBMCKEFDI-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole, isoxazole, 3,5-dimethyl, 3,5-dimethylisooxazole, 3,5-dwumetyloizoksazolu, 3,5-dwumetyloizoksazolu polish, 3,5-dimethylisoxazol, pubchem8641, isoxazole,5-dimethyl, 3,5-dimethyl-isoxazole
PubChem CID 9312
IUPAC Name 3,5-dimethyl-1,2-oxazole
SMILES CC1=CC(=NO1)C

Specifications

Melting Point -14°C
Density 0.992
Boiling Point 142°C to 144°C
Flash Point 31°C (87°F)
Refractive Index 1.4415
Quantity 100 g
UN Number UN1993
Beilstein 106324
Solubility Information Insoluble in water.
Formula Weight 97.12
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylisoxazole
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-100-2
RTECSNumber NY2774200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.