Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,5-Dimethylisoxazole 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
250 mL
Specifications
Specifications
Color | Yellow |
Boiling Point | 141°C |
Quantity | 250 mL |
UN Number | 1993 |
Formula Weight | 97.12 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 3,5-Dimethylisoxazole |
Safety and Handling
RTECSNumber | NY2774200 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction