Learn More
3,5-Dimethylphenol, 98+%
Description
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
Solubility
Soluble in ethyl alcohol, carbon tetrachloride, benzene, chloroform, water and ether.
Notes
Incompatible with bases, acid chlorides, acid anhydrides, oxidizing agents, brass and copper.

Specifications
Specifications
Melting Point | 61°C to 65°C |
Density | 1.12 |
Boiling Point | 222°C |
Flash Point | 109°C (228°F) |
Quantity | 500 g |
UN Number | UN2261 |
Beilstein | 774117 |
Merck Index | 14,10082 |
Solubility Information | Soluble in ethyl alcohol,carbon tetrachloride,benzene,chloroform,water and ether. |
Formula Weight | 122.17 |
Show More |
Safety and Handling
Hazard Category | H301+H311-H314 |
Hazard Statement | GHS H Statement H301-H311-H314 Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. |
Precautionary Statement | P260-P264b-P270-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID |
EINECSNumber | 203-606-5 |
RTECSNumber | ZE6475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.